ADVH49B835F0 AChemBlock

3-chlorobenzofuran

  • Cas Number : 63361-59-1
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chlorobenzofuran.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89155
Availability details for
Quantity
  • Availability
  • Add to Cart