ADVH49B835F1 AChemBlock

ethyl 4-hydroxy-1,8-naphthyridine-3-carboxylate

  • Cas Number : 67992-11-4
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-hydroxy-1-8-naphthyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89156
Availability details for
Quantity
  • Availability
  • Add to Cart