ADVH49B835F4 AChemBlock

2-(3-aminopyridin-2-yl)acetic acid

  • Cas Number : 80352-63-2
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-aminopyridin-2-yl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89159
Availability details for
Quantity
  • Availability
  • Add to Cart