ADVH49B835F6 AChemBlock

(S)-3-((S)-1-oxo-3a,4,5,6-tetrahydro-1H-benzo[de]isoquinolin-2(3H)-yl)quinuclidine 1-oxide

  • Cas Number : 813425-83-1
  • Molecular Weight : 312.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-s-1-oxo-3a-4-5-6-tetrahydro-1h-benzo-de-isoquinolin-2-3h-yl-quinuclidine-1-oxide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89161
Availability details for
Quantity
  • Availability
  • Add to Cart