ADVH49B835F8 AChemBlock

TCS1102

  • Cas Number : 916141-36-1
  • Molecular Weight : 470.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tcs1102.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89163
Availability details for
Quantity
  • Availability
  • Add to Cart