ADVH49B835FB AChemBlock

propane-2,2-diylbis(4,1-phenylene) diacrylate

  • Molecular Weight : 336.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/propane-2-2-diylbis-4-1-phenylene-diacrylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89166
Availability details for
Quantity
  • Availability
  • Add to Cart