ADVH49B835FD AChemBlock

7-(tert-butoxycarbonyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine-3-carboxylic acid

  • Molecular Weight : 268.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-tert-butoxycarbonyl-5-6-7-8-tetrahydro-1-2-4-triazolo-4-3-a-pyrazine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89168
Availability details for
Quantity
  • Availability
  • Add to Cart