ADVH49B83600 AChemBlock

LAZABEMIDE

  • Cas Number : 103878-84-8
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/lazabemide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89171
Availability details for
Quantity
  • Availability
  • Add to Cart