ADVH49B83601 AChemBlock

2-(dimethylamino)-6-fluorobenzamide

  • Cas Number : 107485-44-9
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-dimethylamino-6-fluorobenzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89172
Availability details for
Quantity
  • Availability
  • Add to Cart