ADVH49B83602 AChemBlock

2-morpholinooxazolo[4,5-b]pyridine

  • Cas Number : 113520-20-0
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-morpholinooxazolo-4-5-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89173
Availability details for
Quantity
  • Availability
  • Add to Cart