ADVH49B83604 AChemBlock

mAChr-in-1

  • Cas Number : 119391-56-9
  • Molecular Weight : 488.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/machr-in-1.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89175
Availability details for
Quantity
  • Availability
  • Add to Cart