ADVH49B83605 AChemBlock

ethyl 4-acetyl-1H-pyrrole-2-carboxylate

  • Cas Number : 119647-69-7
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-acetyl-1h-pyrrole-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89176
Availability details for
Quantity
  • Availability
  • Add to Cart