ADVH49B83606 AChemBlock

4-((2-(pyrrolidin-1-yl)ethyl)amino)benzoic acid

  • Cas Number : 1225863-00-2
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-2-pyrrolidin-1-yl-ethyl-amino-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89177
Availability details for
Quantity
  • Availability
  • Add to Cart