ADVH49B8360A AChemBlock

N-(2,4-dichlorobenzyl)hydroxylamine hydrochloride

  • Cas Number : 139460-29-0
  • Molecular Weight : 228.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-2-4-dichlorobenzyl-hydroxylamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89181
Availability details for
Quantity
  • Availability
  • Add to Cart