ADVH49B8360C AChemBlock

1-(4'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl)ethan-1-one

  • Cas Number : 142557-76-4
  • Molecular Weight : 264.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-trifluoromethyl-1-1-biphenyl-4-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89183
Availability details for
Quantity
  • Availability
  • Add to Cart