ADVH49B8360E AChemBlock

2-hydroxy-5-((2-hydroxybenzyl)amino)benzoic acid

  • Cas Number : 151391-87-6
  • Molecular Weight : 259.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-hydroxy-5-2-hydroxybenzyl-amino-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89185
Availability details for
Quantity
  • Availability
  • Add to Cart