ADVH49B8360F AChemBlock

10-methyl-9-(phenoxycarbonyl)acridin-10-ium trifluoromethanesulfonate

  • Cas Number : 161006-14-0
  • Molecular Weight : 463.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/10-methyl-9-phenoxycarbonyl-acridin-10-ium-trifluoromethanesulfonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89186
Availability details for
Quantity
  • Availability
  • Add to Cart