ADVH49B83613 AChemBlock

CA-4948

  • Cas Number : 1801343-74-7
  • Molecular Weight : 307.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ca-4948.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89190
Availability details for
Quantity
  • Availability
  • Add to Cart