ADVH49B83614 AChemBlock

dimethyl 2-(2-(4-chlorophenyl)-2-oxoethyl)malonate

  • Cas Number : 194653-25-3
  • Molecular Weight : 284.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/dimethyl-2-2-4-chlorophenyl-2-oxoethyl-malonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89191
Availability details for
Quantity
  • Availability
  • Add to Cart