ADVH49B83615 AChemBlock

GNE-6640

  • Cas Number : 2009273-67-8
  • Molecular Weight : 330.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/gne-6640.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89192
Availability details for
Quantity
  • Availability
  • Add to Cart