ADVH49B83616 AChemBlock

GET-73

  • Cas Number : 202402-01-5
  • Molecular Weight : 275.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/get-73.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89193
Availability details for
Quantity
  • Availability
  • Add to Cart