ADVH49B8361A AChemBlock

1-(naphthalen-2-yl)-2-(pyridin-4-yl)ethan-1-one

  • Cas Number : 224040-86-2
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-naphthalen-2-yl-2-pyridin-4-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89197
Availability details for
Quantity
  • Availability
  • Add to Cart