ADVH49B8361B AChemBlock

methyl 3-bromo-4-ethoxybenzoate

  • Cas Number : 24507-28-6
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-bromo-4-ethoxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89198
Availability details for
Quantity
  • Availability
  • Add to Cart