ADVH49B8361D AChemBlock

3-cyano-4-ethoxybenzoic acid

  • Cas Number : 258273-32-4
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-cyano-4-ethoxybenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89200
Availability details for
Quantity
  • Availability
  • Add to Cart