ADVH49B83620 AChemBlock

HT-2157

  • Molecular Weight : 366.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/ht-2157.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89203
Availability details for
Quantity
  • Availability
  • Add to Cart