ADVH49B83621 AChemBlock

methyl 3-cyano-4-ethoxybenzoate

  • Cas Number : 318528-53-9
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-cyano-4-ethoxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89204
Availability details for
Quantity
  • Availability
  • Add to Cart