ADVH49B83625 AChemBlock

ZD-4190

  • Cas Number : 413599-62-9
  • Molecular Weight : 495.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/zd-4190.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89208
Availability details for
Quantity
  • Availability
  • Add to Cart