ADVH49B83626 AChemBlock

4-(2H-benzo[d][1,2,3]triazol-2-yl)butanoic acid

  • Cas Number : 4144-70-1
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-2h-benzo-d-1-2-3-triazol-2-yl-butanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89209
Availability details for
Quantity
  • Availability
  • Add to Cart