ADVH49B83629 AChemBlock

2-oxo-2-(2-oxo-2,3-dihydrobenzo[d]oxazol-6-ylamino)acetic acid

  • Cas Number : 496055-50-6
  • Molecular Weight : 222.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-oxo-2-2-oxo-2-3-dihydrobenzo-d-oxazol-6-ylamino-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89212
Availability details for
Quantity
  • Availability
  • Add to Cart