ADVH49B8362A AChemBlock

GUANABENZ

  • Cas Number : 5051-62-7
  • Molecular Weight : 231.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/guanabenz.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89213
Availability details for
Quantity
  • Availability
  • Add to Cart