ADVH49B8362E AChemBlock

N,N-dimethylpropanethioamide

  • Cas Number : 5309-94-4
  • Molecular Weight : 117.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-n-dimethylpropanethioamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89217
Availability details for
Quantity
  • Availability
  • Add to Cart