ADVH49B83630 AChemBlock

4,6-dichloro-N-(4-fluorobenzyl)-1,3,5-triazin-2-amine

  • Cas Number : 546140-76-5
  • Molecular Weight : 273.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-n-4-fluorobenzyl-1-3-5-triazin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89219
Availability details for
Quantity
  • Availability
  • Add to Cart