ADVH49B83632 AChemBlock

6-(naphthalen-2-yl)-5-(pyridin-4-yl)pyridazin-3(2H)-one

  • Cas Number : 629623-78-5
  • Molecular Weight : 299.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-naphthalen-2-yl-5-pyridin-4-yl-pyridazin-3-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89221
Availability details for
Quantity
  • Availability
  • Add to Cart