ADVH49B83634 AChemBlock

ethyl 4-(1H-benzo[d][1,2,3]triazol-1-yl)butanoate

  • Cas Number : 69218-48-0
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-1h-benzo-d-1-2-3-triazol-1-yl-butanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89223
Availability details for
Quantity
  • Availability
  • Add to Cart