ADVH49B83635 AChemBlock

ethyl 4-(2H-benzo[d][1,2,3]triazol-2-yl)butanoate

  • Cas Number : 69218-53-7
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-2h-benzo-d-1-2-3-triazol-2-yl-butanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89224
Availability details for
Quantity
  • Availability
  • Add to Cart