ADVH49B83636 AChemBlock

Tyrosine kinase-IN-1

  • Cas Number : 705946-27-6
  • Molecular Weight : 445.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tyrosine-kinase-in-1.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89225
Availability details for
Quantity
  • Availability
  • Add to Cart