ADVH49B83637 AChemBlock

methyl (naphthalen-2-ylmethyl) carbonate

  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-naphthalen-2-ylmethyl-carbonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88657
Availability details for
Quantity
  • Availability
  • Add to Cart