ADVH49B8363D AChemBlock

AZD 3241

  • Cas Number : 890655-80-8
  • Molecular Weight : 253.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/azd-3241.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89232
Availability details for
Quantity
  • Availability
  • Add to Cart