ADVH49B83641 AChemBlock

2-(methylthio)pyrido[3,4-d]pyrimidin-4(3H)-one

  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methylthio-pyrido-3-4-d-pyrimidin-4-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89236
Availability details for
Quantity
  • Availability
  • Add to Cart