ADVH49B83642 AChemBlock

1-isopropyl-1H-tetrazole

  • Cas Number : 100114-32-7
  • Molecular Weight : 112.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-isopropyl-1h-tetrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89237
Availability details for
Quantity
  • Availability
  • Add to Cart