ADVH49B83646 AChemBlock

6-bromo-1-nitronaphthalene

  • Cas Number : 102153-48-0
  • Molecular Weight : 252.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-1-nitronaphthalene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89241
Availability details for
Quantity
  • Availability
  • Add to Cart