ADVH49B83648 AChemBlock

5-(trifluoromethyl)-1H-imidazole-2-carbaldehyde

  • Cas Number : 102808-02-6
  • Molecular Weight : 164.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-1h-imidazole-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89243
Availability details for
Quantity
  • Availability
  • Add to Cart