ADVH49B83649 AChemBlock

methyl 4-iodo-2-methylbenzoate

  • Cas Number : 103440-53-5
  • Molecular Weight : 276.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-iodo-2-methylbenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89244
Availability details for
Quantity
  • Availability
  • Add to Cart