ADVH49B8364D AChemBlock

methyl 2-(2-aminoethyl)benzoate hydrochloride

  • Cas Number : 1046758-68-2
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-2-aminoethyl-benzoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89248
Availability details for
Quantity
  • Availability
  • Add to Cart