ADVH49B8364E AChemBlock

quinoline-5,8-dione

  • Cas Number : 10470-83-4
  • Molecular Weight : 159.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/quinoline-5-8-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89249
Availability details for
Quantity
  • Availability
  • Add to Cart