ADVH49B8364F AChemBlock

methyl 4-bromo-5-chlorothiophene-2-carboxylate

  • Cas Number : 1047630-72-7
  • Molecular Weight : 255.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-5-chlorothiophene-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89250
Availability details for
Quantity
  • Availability
  • Add to Cart