ADVH49B83651 AChemBlock

1-(3,4-dimethoxyphenyl)-3-hydroxy-2-(2-methoxyphenoxy)propan-1-one

  • Cas Number : 10548-77-3
  • Molecular Weight : 332.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-3-4-dimethoxyphenyl-3-hydroxy-2-2-methoxyphenoxy-propan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89252
Availability details for
Quantity
  • Availability
  • Add to Cart