ADVH49B83653 AChemBlock

hexyl 3,4,5-trihydroxybenzoate

  • Cas Number : 1087-26-9
  • Molecular Weight : 254.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/hexyl-3-4-5-trihydroxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89254
Availability details for
Quantity
  • Availability
  • Add to Cart