ADVH49B83656 AChemBlock

3,4-dibromofuran-2,5-dione

  • Cas Number : 1122-12-9
  • Molecular Weight : 255.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-4-dibromofuran-2-5-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89257
Availability details for
Quantity
  • Availability
  • Add to Cart