ADVH49B8365E AChemBlock

5-(4'-methyl-[1,1'-biphenyl]-2-yl)-1H-tetrazole

  • Cas Number : 120568-11-8
  • Molecular Weight : 236.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-4-methyl-1-1-biphenyl-2-yl-1h-tetrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89265
Availability details for
Quantity
  • Availability
  • Add to Cart